missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM65B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548820
This item is not returnable.
View return policy
Description
FAM65B Polyclonal Antibody for Western Blot
Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
Specifications
| FAM65B | |
| Polyclonal | |
| Unconjugated | |
| Ripor2 | |
| Ab2-162; C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; family with sequence similarity 65 member B; family with sequence similarity 65, member B; Kiaa0386; myogenesis-related and NCAM-associated protein homolog; MYONAP; PL48; protein FAM65B; RGD1306939; Rho family-interacting cell polarization regulator 2; RIPOR2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9Y4F9 | |
| Ripor2 | |
| KLH conjugated synthetic peptide between 829-856 amino acids from the C-terminal region of human FAM65B. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction