missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM65B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 483.00
Specifications
| Antigen | FAM65B |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315976
|
Novus Biologicals
NBP3-04522-20UL |
20 μL |
€ 204.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017233
|
Novus Biologicals
NBP3-04522-100UL |
100 μL |
€ 483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM65B Polyclonal specifically detects FAM65B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| FAM65B | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 9750 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C6orf32, DIFF48DIFF40, family with sequence similarity 65, member B, PL48KIAA0386chromosome 6 open reading frame 32 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human FAM65B (NP_056948.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title