missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAM70B Antibody [Janelia Fluor™ 646], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30755JF646
This item is not returnable.
View return policy
Description
FAM70B Polyclonal antibody specifically detects FAM70B in Human samples. It is validated for Western Blot
Specifications
| FAM70B | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 348013 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| family with sequence similarity 70, member B, FLJ57938, MGC20579, RP11-199F6.1 | |
| Produced in rabbits immunized with E. coli-derived Human FAM70B fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction