missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM78A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21038-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM78A is a protein coding gene.Specifications
| FAM78A | |
| Polyclonal | |
| Unconjugated | |
| FAM78A | |
| C9orf59, FAM78A, Protein FAM78A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 241303, 286336, 499776 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JUQ0, Q8C552 | |
| FAM78A | |
| FAM78A Fusion Protein Ag15366 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction