missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM82A Antibody, DyLight 488, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41121G
This item is not returnable.
View return policy
Description
FAM82A Polyclonal antibody specifically detects FAM82A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| FAM82A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 151393 | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 488 | |
| family with sequence similarity 82, member A1, member A, UNQ9371/PRO34163 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM82A1. The immunogen is located within amino acids 140 - 190 of FAM82A1. Amino Acid Squence: ITANTDTEEQSFPVPK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction