missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM83H Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32219CL1
This item is not returnable.
View return policy
Description
FAM83H Polyclonal antibody specifically detects FAM83H in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| FAM83H | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 286077 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| family with sequence similarity 83, member H, protein FAM83H | |
| The immunogen this antibody was made to, maps to a region between residue 1129 to 1179 of human Family with Sequence Similarity 83, Member H using the numbering given in entry NP_940890.3 (GeneID 286077). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction