missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM84B Antibody (OTI1D12), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72440PE
This item is not returnable.
View return policy
Description
FAM84B Monoclonal specifically detects FAM84B in Human samples. It is validated for Flow Cytometry.
Specifications
| FAM84B | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| FAM84B | |
| Full length recombinant protein of human FAM84B (NP_777571) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1D12 | |
| Flow Cytometry | |
| BCMP101neurological/sensory 2, Breast cancer membrane protein 101, breast cancer membrane-associated protein 101, family with sequence similarity 84, member B, NSE2Protein NSE2 | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 157638.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction