missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM8A1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47125DL594
This item is not returnable.
View return policy
Description
FAM8A1 Polyclonal antibody specifically detects FAM8A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FAM8A1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51439 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| AHCP | |
| A synthetic peptide made to an internal portion of the human FAM8A1 protein (between amino acids 50-100) [Uniprot Q9UBU6] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction