missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM92A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584396
This item is not returnable.
View return policy
Description
Immunogen sequence: VERLEAKVVE PLKTYGTIVK MKRDDLKATL TARNREAKQL TQLERTRQRN PSDRHVISQA ETELQRAAMD ASRTSRHLE Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 94%.
Acts as a positive regulator of ciliary hedgehog signaling. Probable regulator of ciliogenesis involved in limb morphogenesis. In cooperation with CBY1 it is involved in the recruitment and fusion of endosomal vesicles at distal appendages during early stages of ciliogenesis. [UniProt].
Specifications
| FAM92A1 | |
| Polyclonal | |
| Unconjugated | |
| FAM92A | |
| 6720467C03Rik; Fam92a; FAM92A1; family with sequence similarity 92 member A1; family with sequence similarity 92, member A; family with sequence similarity 92, member A1; Protein FAM92A; protein FAM92A1; RGD1310681 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 137392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1XBS5 | |
| FAM92A | |
| Recombinant protein corresponding to Human FAM92A. Recombinant protein control fragment (Product #RP-108214). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction