missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM98A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | FAM98A |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227319
|
Novus Biologicals
NBP3-35605-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232141
|
Novus Biologicals
NBP3-35605-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM98A Polyclonal antibody specifically detects FAM98A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| FAM98A | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZP564F0522, DKFZp686O03192, family with sequence similarity 98, member A, hypothetical protein LOC25940 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human FAM98A (NP_056290.3).,, Sequence:, MECDLMETDILESLEDLGYKGPLLEDGALSQAVSAGASSPEFTKLCAWLVSELRVLCKLEENVQATNSPSEAEEFQLEVSGLLGEMNCPYLSLTSGDVTKRLLIQKNCLLLLTYLISELEAARMLCVNAPPKKAQEGGGSEVFQELKGICIALGMSKPPANITMFQFFSGIEKKLKETLAKVPPNHVGKPLLKKPMGPAHWEKIEAINQAIANEYEVRRK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 25940 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title