missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM98B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552568
This item is not returnable.
View return policy
Description
Immunogen sequence: GPQPTMEGDV LDTLEALGYK GPLLEEQALT KAAEGGLSSP EFSELCIWLG SQIKSLCNLE ESITSAGRDD LESFQLEISG FLKEMACPYS VLISGDIKDR LKKKEDCLKL LLFLSTELQA SQILQN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
FAM98B positively stimulates PRMT1-induced protein arginine dimethylated arginine methylation. Promotes colorectal cancer cell malignancy.
Specifications
| FAM98B | |
| Polyclonal | |
| Unconjugated | |
| FAM98B | |
| 2610510H03Rik; Fam98b; family with sequence similarity 98 member B; family with sequence similarity 98, member B; protein FAM98B; RGD1564603 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283742, 499866, 68215 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q52LJ0, Q80VD1 | |
| FAM98B | |
| Recombinant protein corresponding to Human FAM98B. Recombinant protein control fragment (Product #RP-89733). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction