Learn More
Famotidine Formaldehyde Adduct Maleate (2-[4-[2-(1,1-Dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate), Mikromol™
Product Code. 17910976
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910976 100 mg 100mg
17964156 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17910976 Supplier Mikromol Supplier No. MM0029.17
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Famotidine Formaldehyde Adduct Maleate (2-[4-[2-(1,1-Dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Famotidine Formaldehyde Adduct Maleate
CAS 2575516-45-7
Analyte or Component Names Famotidine Formaldehyde Adduct Maleate
Shipping Condition Room Temperature
Molecular Formula C9H15N7O2S3.C4H4O4
Certifications/Compliance ISO 9001
Synonym [4-[[[2-(3,4-dihydro-1,1-dioxido-2H-1,2,4,6-thiatriazin-3-yl)ethyl]thio]methyl]-2-thiazolyl]guanidine Maleate, 2-(4-(((2-(1,1-Dioxido-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethyl)sulfanyl)methyl)-1,3-thiazol-2-yl)guanidine maleate, Famotidine Impurity C1 as Maleate, Famotidine Formaldehyde Adduct Maleate
InChI Formula InChI=1S/C9H15N7O2S3.C4H4O4/c10-8(11)15-9-14-6(4-20-9)3-19-2-1-7-12-5-13-21(17,18)16-7;5-3(6)1-2-4(7)8/h4,13H,1-3,5H2,(H,12,16)(H4,10,11,14,15);1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES NC(=Nc1nc(CSCCC2=NS(=O)(=O)NCN2)cs1)N.OC(=O)\C=C/C(=O)O
IUPAC Name 2-[4-[2-(1,1-dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]thiazol-2-yl]guanidine;maleic acid
Molecular Weight (g/mol) 465.53
Quantity 100 mg
Formula Weight 465.0559 g/mol