Learn More
Binder™ Fan Replacement Set (230 V)
Shop All BINDER Products
Click to view available options
Description:
Fan Replacement Set (230V)
Unit Size:
Each
Description
Fan replacement set for FD/FED with forced convection
- Fan replacement set for FD/FED with forced convection
Specifications
Specifications
| Description | Fan Replacement Set (230V) |
| For Use With (Equipment) | BINDER Chambers FD 56, FD 115, FD 260, FD 720 |
| Type | Fan Motor |
| Voltage | 230 V |