Learn More
Memmert™ Fan Speed Monitoring for IF160plus
Product Code. 17586775 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF160plus Incubator
Unit Size:
Each
Product Code. 17586775
Brand: Memmert™ IF160plusV4
Only null left
Add to basket
Add to basket
Fan speed monitoring with switch-off the heating feature and alarm in case of failure.
  • With switch-off heating
  • With alarm
Specifications
For Use With (Equipment) IF160plus Incubator
Type Fan Speed Monitor
Includes Switch and alarm