Learn More
Memmert™ Fan Speed Monitoring for IF450plus
Product Code. 17557055 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IF450plus Incubator
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17557055 IF450plus Incubator Each
1 options
Product Code. 17557055
Brand: Memmert™ IF450plusV4
Only null left
Add to basket
Add to basket
Fan speed monitoring with switch-off the heating feature and alarm in case of failure.
  • With switch-off heating
  • With alarm
Specifications
For Use With (Equipment) IF450plus Incubator
Type Fan Speed Monitor
Includes Switch and alarm