missing translation for 'onlineSavingsMsg'
Learn More

FANCA 293T Cell Overexpression Lysate (Denatured), Abnova

Product Code. 16194821
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 16194821

Brand: Abnova H00002175T01.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell transient overexpression lysate for FANCA

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2).

  • Fanconi anemia, complementation group A
  • Transfected cell line: 293T
  • Plasmid: pCMV-FANCA full-length
  • Host: Human
  • Theoretical molecular weight: 32.78kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description FANCA 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC008979
Molecular Weight (g/mol) 32.78
pH Range 6.8
Plasmid pCMV-FANCA full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-FANCA antibody (H00002175-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL FANCA 293T Cell Transient Overexpression Lys
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.