missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCB Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19087JF646
This item is not returnable.
View return policy
Description
FANCB Polyclonal antibody specifically detects FANCB in Human samples. It is validated for Western Blot
Specifications
| FANCB | |
| Polyclonal | |
| Western Blot | |
| EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB | |
| The immunogen recognized by this antibody maps to a region between residue 820 and 859 of human Fanconi anemia complementation group B using the numbering given in entry NP_001018123.1 (GeneID 2187). | |
| 0.1 mL | |
| Cancer, DNA Repair, Epigenetics | |
| 2187 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction