Learn More
FANCD2 Antibody (1290D), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB9369E
Product Code. 30066228
  • € 629.00 / 0.10mL
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FANCD2
Monoclonal
DyLight 405
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Flow Cytometry, KnockDown
1290D
Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
E. coli-derived recombinant human FANCD2, Glu11-Glu230, Accession # Q9BXW9
0.1 mL
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
2177
Store at 4C in the dark.