missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (1290D) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9369JF635
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Gene Knock-Out,Dual RNAscope ISH-IHC
Specifications
| FANCD2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| E. coli-derived recombinant human FANCD2, Glu11-Glu230, Accession # Q9BXW9 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Dual RNAScope ISH-IHC | |
| 1290D | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction