missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCD2 Polyclonal Antibody, DyLight™ 550

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522708
This item is not returnable.
View return policy
Description
In Western blot, this antibody should recognize a band at ~166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights. In immunofluorescence, this has been tested in human MMC and IR-treated MEF cells.
FANCD2 (Fanconi anemia group D2 protein) is required for maintenance of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.
Specifications
| FANCD2 | |
| Polyclonal | |
| DyLight 550 | |
| FANCD2 | |
| 2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 211651, 2177, 312641, 394241, 484659 | |
| 4°C, store in dark | |
| Liquid |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| Q6IV68, Q80V62, Q9BXW9 | |
| FANCD2 | |
| Human FANCD2 fusion protein (N-terminal fragment). | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Avian, Zebrafish, Marsupial | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction