missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576383
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group E.
Specifications
| FANCE | |
| Polyclonal | |
| Unconjugated | |
| FANCE | |
| 2810451D06Rik; AI415634; AW209126; FACE; FAE; FANCE; Fanconi anemia complementation group E; Fanconi anemia group E protein; Fanconi anemia, complementation group E; Protein FACE; RGD1561045 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2178, 72775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HB96 | |
| FANCE | |
| Recombinant fusion protein of human FANCE. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction