missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | FANCI |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228360
|
Novus Biologicals
NBP3-38040-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231524
|
Novus Biologicals
NBP3-38040-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for ELISA,Western BlotSpecifications
| FANCI | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 55215 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1061-1328 of human FANCI (NP_001106849.1).,, Sequence:, DQDVEVEKTNHFAIVNLRTAAPTVCLLVLSQAEKVLEEVDWLITKLKGQVSQETLSEEASSQATLPNQPVEKAIIMQLGTLLTFFHELVQTALPSGSCVDTLLKDLCKMYTTLTALVRYYLQVCQSSGGIPKNMEKLVKLSGSHLTPLCYSFISYVQNKSKSLNYTGEKKEKPAAVATAMARVLRETKPIPNLIFAIEQYEKFLIHLSKKSKVNLMQHMKLSTSRDFKIKGNILDMVLREDGEDENEEGTASEHGGQNKEPAKKKRKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title