missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FANK1 Polyclonal Antibody

Product Code. 15898173
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15898173 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898173 Supplier Invitrogen™ Supplier No. PA569769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.

Through the activation of JUN and AP-1-mediated transcription, may regulate apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANK1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FANK1
Gene Accession No. Q9DAM9
Gene Alias 1700007B22Rik; AI850911; FANK1; fibronectin type 3 and ankyrin repeat domains 1; fibronectin type 3 and ankyrin repeat domains 1 protein; Fibronectin type 3 and ankyrin repeat domains protein 1; fibronectin type III and ankyrin repeat domains 1; germ cell-specific 1; germ cell-specific gene 1 protein; GSG1; GV14; HSD13; UNQ6504/PRO21382
Gene Symbols FANK1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse FANK1 (aa 125-174).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66930
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.