Learn More
Hellma™ Far UV Quartz Fluorescence Cuvette Single Cells
Click to view available options
Capacity (Metric):
0.1 mL
Unit Size:
Each
Specifications
Specifications
| Type | Fluorescence Cuvette Single Cell |
| Capacity (Metric) | 0.1 mL |
| Material | Far UV Quartz |
| Quantity | 1Each |
| Path Length | 10 mm |
| Wavelength Range | 200 to 2500 nm |