missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FARSB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557764
This item is not returnable.
View return policy
Description
Immunogen sequence: THDLDTLSGP FTYTAKRPSD IKFKPLNKTK EYTACELMNI YKTDNHLKHY LHIIENKPLY PVIYDSNGVV LSMPPIINGD HSRITVNT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes a highly conserved enzyme that belongs to the aminoacyl-tRNA synthetase class IIc subfamily. This enzyme comprises the regulatory beta subunits that form a tetramer with two catalytic alpha subunits. In the presence of ATP, this tetramer is responsible for attaching L-phenylalanine to the terminal adenosine of the appropriate tRNA. A pseudogene located on chromosome 10 has been identified.
Specifications
| FARSB | |
| Polyclonal | |
| Unconjugated | |
| Farsb | |
| C76708; Farsa; FARSB; Farsl; Farslb; Frsb; HSPC173; ph; PheHB; phenylalanine tRNA ligase 1, beta, cytoplasmic; phenylalanine tRNA synthetase alpha; phenylalanine-tRNA ligase beta chain; phenylalanine--tRNA ligase beta chain; phenylalanine--tRNA ligase beta subunit; phenylalanine-tRNA synthetase-like, beta subunit; phenylalanyl-tRNA synthetase beta chain; phenylalanyl-tRNA synthetase beta subunit; phenylalanyl-tRNA synthetase beta-subunit; phenylalanyl-tRNA synthetase, beta subunit; phenylalanyl-tRNA synthetase-like, beta subunit; PheRS; PheRS alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NSD9 | |
| Farsb | |
| Recombinant protein corresponding to Human FARSB. Recombinant protein control fragment (Product #RP-96043). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction