Learn More
FARSLA Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97761PCP
Product Code. 18614922
  • € 407.00 / 0.10mL
Description
FARSLA Polyclonal antibody specifically detects FARSLA in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PerCP
RUO
Primary
0.1 mL
Core ESC Like Genes, Stem Cell Markers
Store at 4C in the dark.