missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas/TNFRSF6/CD95 Antibody (50825), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB326F
This item is not returnable.
View return policy
Description
Fas/TNFRSF6/CD95 Monoclonal antibody specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Flow Cytometry
Specifications
| Fas/TNFRSF6/CD95 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 50825 | |
| Flow Cytometry | |
| Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
| Mouse myeloma cell line NS0-derived recombinant human Fas/TNFRSF6/CD95, Gln26-Asn173, Accession # P25445 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells | |
| 355 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction