missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas/TNFRSF6/CD95 Rabbit anti-Rat, Clone: 28, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 552.00
Specifications
| Antigen | Fas/TNFRSF6/CD95 |
|---|---|
| Clone | 28 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Rat samples. It is validated for Flow Cytometry.Specifications
| Fas/TNFRSF6/CD95 | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 355 | |
| IgG | |
| Protein A purified |
| 28 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6 | |
| Recombinant Rat Fas/TNFRSF6/CD95 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title