Learn More
Fascin Antibody (FSCN1/416), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-11582CL1
Product Code. 30147559
  • € 1100.00 / 0.10mL
Description
Fascin Monoclonal antibody specifically detects Fascin in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fascin
Monoclonal
CoraFluor 1
FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin
Full length recombinant human Fascin protein (Uniprot: Q16658)
0.1 mL
Cell Biology, Cellular Markers
6624
Store at 4°C in the dark. Do not freeze.
IgG2a κ
Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
FSCN1/416