Learn More
Fascin Antibody (FSCN1/417), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47801MFV500
Product Code. 18717532
  • € 735.00 / 0.10mL
Description
Fascin Monoclonal antibody specifically detects Fascin in Human samples. It is validated for Flow Cytometry