missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10624-2-AP-20UL
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.Specifications
| FASN | |
| Polyclonal | |
| Unconjugated | |
| FASN | |
| FAS, FASN, fatty acid synthase, OA 519, SDR27X1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14104, 2194, 50671 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Flow Cytometry | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12785, P19096, P49327 | |
| Fasn | |
| FASN Fusion Protein Ag0975 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction