missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32275R
This item is not returnable.
View return policy
Description
FAT1 Polyclonal specifically detects FAT1 in Human samples. It is validated for Western Blot.
Specifications
| FAT1 | |
| Polyclonal | |
| Western Blot | |
| FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FAT1 | |
| The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195). | |
| 100 μL | |
| Signal Transduction | |
| 2195 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction