missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT4 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18503JF669
This item is not returnable.
View return policy
Description
FAT4 Polyclonal antibody specifically detects FAT4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| FAT4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 669 | |
| cadherin family member 14, cadherin-related family member 11, CDHR11, FAT tumor suppressor homolog 4 (Drosophila), FAT-J, fat-like cadherin protein FAT-J, FLJ77878, hFat4, Nbla00548, putative protein product of Nbla00548 | |
| Recognizes region between residues Between between 4931 and 4981.. | |
| 0.1 mL | |
| Cancer, Cell Biology, Neuroscience | |
| 79633 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction