missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATE1 Monoclonal Antibody (OTI5E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505183
This item is not returnable.
View return policy
Description
This gene encodes a cancer-testis antigen that is highly expressed in hepatocellular carcinomas and other tumors and weakly expressed in normal tissues except testis. The protein is strongly expressed in spermatogonia, primary spermatocytes, and Sertoli cells in seminiferous tubules. This protein may have a role in the control of early testicular differentiation and cell proliferation.Specifications
| FATE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969F0 | |
| FATE1 | |
| Full length human recombit protein of human FATE1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5E2 | |
| Unconjugated | |
| FATE1 | |
| CT43, FATE | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction