missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FATE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15724891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15724891 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15724891 Supplier Invitrogen™ Supplier No. PA556989

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GSQLPKPRML RESGHGDAHL QEYAGNFQGI RFHYDRNPGT DAVAQTSLEE FNVLEMEVMR RQLYAVNRRL RALEEQGATW Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 26%.

This gene encodes a cancer-testis antigen that is highly expressed in hepatocellular carcinomas and other tumors and weakly expressed in normal tissues except testis. The protein is strongly expressed in spermatogonia, primary spermatocytes, and Sertoli cells in seminiferous tubules. This protein may have a role in the control of early testicular differentiation and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FATE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FATE1
Gene Accession No. Q969F0
Gene Alias 9230118O15Rik; BJ-HCC-2 antigen; cancer/testis antigen 43; CT43; EGK_21049; FATE; FATE1; fetal and adult testis expressed 1; fetal and adult testis expressed transcript protein; fetal and adult testis-expressed transcript protein; Tumor antigen BJ-HCC-2
Gene Symbols FATE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FATE1. Recombinant protein control fragment (Product #RP-95451).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89885
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.