Learn More
FATP4/SLC27A4 Antibody (11A1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26320-50ul
 View more versions of this product
Product Code. 30037258
  • € 343.00 / 50µL
Description
FATP4/SLC27A4 Monoclonal antibody specifically detects FATP4/SLC27A4 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FATP4/SLC27A4
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
10999
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA
11A1
Western Blot 1:500-1:2000, ELISA
ACSVL4Solute carrier family 27 member 4, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-4, FATP4Fatty acid transport protein 4, IPS, long-chain fatty acid transport protein 4, solute carrier family 27 (fatty acid transporter), member 4
A synthesized peptide derived from Human FATP4/SLC27A4 [UniProt Q6P1M0]
50 μL
Primary
Human, Mouse