missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBLIM1, Mouse anti-Human, Clone: 3F8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00054751-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: VTCARCIGDESFALGSQNEVYCLDDFYRKFAPVCSICENPIIPRDGKDAFKIECMGRNFHENCYRCEDCRILLSVEPTDQGCYPLNNHLFCKPCHVKRSAAGCCSpecifications
| FBLIM1 | |
| Monoclonal | |
| Unconjugated | |
| filamin binding LIM protein 1 | |
| CAL/DKFZp434G171/FBLP-1/FBLP1/RP11-169K16.5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3F8 | |
| In 1x PBS, pH 7.4 | |
| NM_017556 | |
| FBLIM1 | |
| FBLIM1 (NP_060026, 270 a.a. ∼ 373 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 54751 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction