missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBP1 Recombinant Rabbit Monoclonal Antibody (JE48-70)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536138
This item is not returnable.
View return policy
Description
FBP1 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
Specifications
| FBP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P09467, P19112, Q9QXD6 | |
| FBP1 | |
| Recombinant protein within Human FBP1 aa 244-338. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE48-70 | |
| Unconjugated | |
| FBP1 | |
| D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; Fbp; FBP 1; FBP1; Fbp2; Fbp-2; Fbp3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fdp; fructose bisphosphatase 1; fructose-1; fructose-1,6- biphosphatase 1; fructose-1,6-bisphosphatase; fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; fructose-bisphosphatase 1; growth-inhibiting protein 17; liver FBPase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14121, 2203, 24362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?