missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBPase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97978-100ul
This item is not returnable.
View return policy
Description
FBPase 1 Polyclonal antibody specifically detects FBPase 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FBPase 1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| D-fructose-1,6-bisphosphate 1-phosphohydrolase 1, EC 3.1.3, EC 3.1.3.11, FBP, FBPase 1, fructose-1,6-bisphosphatase 1, fructose-bisphosphatase 1, growth-inhibiting protein 17 | |
| Produced in rabbits immunized with E. coli-derived Human FBPase 1 fragment. | |
| 100 μg | |
| Lipid and Metabolism, Protein Phosphatase | |
| 2203 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction