missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBX09 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21448UV
This item is not returnable.
View return policy
Description
FBX09 Polyclonal antibody specifically detects FBX09 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FBX09 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1 | |
| Produced in rabbits immunized with E. coli-derived Human FBX09 fragment | |
| 0.1 mL | |
| Stem Cell Markers | |
| 26268 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction