missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL16 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76527AF700
This item is not returnable.
View return policy
Description
FBXL16 Polyclonal antibody specifically detects FBXL16 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FBXL16 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| C16orf22, c380A1.1, c380A1.1 (novel protein), chromosome 16 open reading frame 22, FBL16, F-box and leucine-rich repeat protein 16Fbl16, F-box/LRR-repeat protein 16, FLJ33735, MGC33974 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human FBXL16. The immunogen is located within the last 50 amino acids of FBXL16. Amino Acid Squence: PGATPELFKYFSQHLPR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 146330 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction