Learn More
FBXL19 Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18015255 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015255 20 μL 20µL
18037265 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18015255 Supplier Proteintech Supplier No. 236241AP20UL
Rabbit Polyclonal Antibody
The FBXL19 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FBXL19. This antibody recognizes human antigen. The FBXL19 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen FBXL19
Applications Western Blot, ELISA
Concentration 550 μg/mL
Gene Symbols F box/LRR repeat protein 19, Fbl19, FBXL19, JHDM1C
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 54620
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG