missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL22 Antibody (1B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00283807-M01
This item is not returnable.
View return policy
Description
FBXL22 Monoclonal antibody specifically detects FBXL22 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| FBXL22 | |
| Monoclonal | |
| Unconjugated | |
| NP_976307 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 283807 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 1B6 | |
| In 1x PBS, pH 7.4 | |
| Fbl22, F-box and leucine-rich repeat protein 22FLJ39626, F-box/LRR-repeat protein 22, MGC75496 | |
| FBXL22 (NP_976307, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MHITQLNRECLLHLFSFLDKDSRKSLARTCSQLHDVFEDPALWSLLHFRSLTELQKDNFLLGPALRSLSICWHSSRVQVCSIEDWLKSAFQRSICSRHESLVNDFLLRVC | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction