Learn More
FBXO11 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98996MFV450
This item is not returnable.
View return policy
Description
FBXO11 Polyclonal antibody specifically detects FBXO11 in Human samples. It is validated for Western Blot
Specifications
| 50mM Sodium Borate | |
| F-box only protein 11, F-box protein 11, FBX11MGC44383, FLJ12673, PRMT9VIT1protein arginine N-methyltransferase 9, ubiquitin protein ligase E3 component n-recognin 6, UBR6, VIT-1, Vitiligo-associated protein 1, vitiligo-associated protein VIT-1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FBXO11. | |
| Purified |