missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO11 Antibody (OTI1F6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72219H
This item is not returnable.
View return policy
Description
FBXO11 Monoclonal specifically detects FBXO11 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FBXO11 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| FBXO11 | |
| Recombinant protein fragment corresponding to amino acids 1-300 of human FBXO11 (NP_079409) produced in E.coli. | |
| 0.1 mL | |
| 80204.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1F6 | |
| Western Blot | |
| F-box only protein 11, F-box protein 11, FBX11MGC44383, FLJ12673, PRMT9VIT1protein arginine N-methyltransferase 9, ubiquitin protein ligase E3 component n-recognin 6, UBR6, VIT-1, Vitiligo-associated protein 1, vitiligo-associated protein VIT-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction