missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO11 Monoclonal Antibody (OTI1F6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526533
This item is not returnable.
View return policy
Description
FBXO11 Monoclonal Antibody for Western Blot
FBXO11 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for FBXO11. This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| FBXO11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86XK2 | |
| FBXO11 | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of FBXO11 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1F6 | |
| Unconjugated | |
| FBXO11 | |
| C80048; F-box only protein 11; F-box protein 11; FBX11; Fbxo11; FLJ12673; GENA 104; jeff; Jf; LOW QUALITY PROTEIN: F-box only protein 11; MGC44383; PRMT9; Protein arginine N-methyltransferase 9; ubiquitin protein ligase E3 component n-recognin 6; UBR6; UG063H01; VIT1; VIT-1; vitiligo-associated protein 1; vitiligo-associated protein VIT-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80204 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction