missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXO11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16529361
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16529361 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16529361 Supplier Invitrogen™ Supplier No. PA596521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GILVYNSGLG CIEDNEIFDN AMAGVWIKTD SNPTLRRNKI HDGRDGGICI FNGGRGLLEE NDIFRNAQAG VLISTNSHPI LRKNRIFDGF AAGIEITNHA TATLEGNQIF NNRFGGLFLA SGVNVTMKDN KIMNNQDAIE KAVSRGQCLY KISSYTSYPM HDFYRCHTCN TTDRNAICVN CIKKCHQGHD VEFIRHDRFF CDCGAGTLSN PCTLAGEPTH DTDTLYDSAP PIESNTLQHN; Positive Samples: HeLa, Mouse spleen, Mouse ovary, Mouse lung; Cellular Location: Chromosome, Nucleus.

FBXO11 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for FBXO11. This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBXO11
Gene Accession No. Q7TPD1, Q86XK2
Gene Alias C80048; F-box only protein 11; F-box protein 11; FBX11; Fbxo11; FLJ12673; GENA 104; jeff; Jf; LOW QUALITY PROTEIN: F-box only protein 11; MGC44383; PRMT9; Protein arginine N-methyltransferase 9; ubiquitin protein ligase E3 component n-recognin 6; UBR6; UG063H01; VIT1; VIT-1; vitiligo-associated protein 1; vitiligo-associated protein VIT-1
Gene Symbols FBXO11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 688-927 of human FBXO11 (NP_0011772031).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225055, 80204
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.