Learn More
FBXO17 Mouse, anti-Human, CoraLite Plus 488, Clone: 1F3D3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68246-100UL
 View more versions of this product
Product Code. 18019495
  • € 450.00 / 100µL
Description
The FBXO17 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO17. This antibody recognizes human antigen. The FBXO17 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
FBXO17
1F3D3
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
F box only protein 17, F box only protein 26, F box protein 17, FBG4, Fbx17, FBX26, FBXO17, FBXO26, FLJ11798, FLJ25205, FLJ58300
100 μL
115290
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.