missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO21 Monoclonal Antibody (OTI5F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504068
This item is not returnable.
View return policy
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants.Specifications
| FBXO21 | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O94952 | |
| FBXO21 | |
| Full length human recombit protein of human FBXO21 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F11 | |
| Unconjugated | |
| FBXO21 | |
| FBX21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23014 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction