missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106917
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FBXO32.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and contains an F-box domain. This protein is highly expressed during muscle atrophy, whereas mice deficient in this gene were found to be resistant to atrophy. This protein is thus a potential drug target for the treatment of muscle atrophy. Alternative splicing of this gene results in two transcript variants encoding two isoforms of different sizes.
Specifications
| FBXO32 | |
| Polyclonal | |
| Unconjugated | |
| FBXO32 | |
| 4833442G10Rik; AI430017; atrogin 1; atrogin1; atrogin-1; atrophy gene 1; F-box only protein 32; F-box protein 32; Fbx32; fbxo25; FBXO32; FLJ32424; Gm20361; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F-box protein; similar to F-box only protein 32 isoform 1; zgc:56479 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114907, 171043, 67731 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91Z62, Q969P5, Q9CPU7 | |
| FBXO32 | |
| A synthesized peptide derived from human FBXO32(Accession Q969P5), corresponding to amino acid residues N50-L100. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction